Learn More
Sino Biological Recombinant Mouse IL-1 beta Protein, HPLC-verified
Supplier:  Sino Biological 50101MNAE1
Encompass_Preferred
A DNA sequence encoding the mature form of mouse IL1β (P10749) (Val 118-Ser 269) was expressed, with an initial Met at the N-terminus.
Catalog No. 50-162-1349