Learn More
ACROBiosystems Recombinant Protein;Human CEACAM-1 / CD66a Protein;HEK293;100UG;CE1,CEACAM1,CD66a,BGP,BGP1,BGP-1,BGPI
Catalog No. 501017513
Encompass_Preferred
Recombinant Protein;Human CEACAM-1 / CD66a Protein;100UG;Human CEACAM-1, His Tag (CE1-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gly 428 (Accession # AAH14473).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 44.2 kDa. The protein migrates as 60-130 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized Human CEACAM-1, His Tag (Cat. No. CE1-H5220) at 10 μg/mL (100 μL/well) can bind Biotinylated Human CEACAM-8, His Tag, Avi Tag (Cat. No. CE8-H82E9) with a linear range of 0.039-1.25 μg/mL (QC tested).
Catalog No. 50-101-7513 Supplier ACROBiosystems Supplier No. CE1H5220100UG
Recombinant Protein;Human CEACAM-1 / CD66a Protein;100UG;Human CEACAM-1, His Tag (CE1-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gly 428 (Accession # AAH14473).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 44.2 kDa. The protein migrates as 60-130 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized Human CEACAM-1, His Tag (Cat. No. CE1-H5220) at 10 μg/mL (100 μL/well) can bind Biotinylated Human CEACAM-8, His Tag, Avi Tag (Cat. No. CE8-H82E9) with a linear range of 0.039-1.25 μg/mL (QC tested).