Learn More
ACROBiosystems Recombinant Protein;Human CD200 / OX-2 Protein;HEK293;100UG;OX2,CD200,MOX1,MOX2,MRC,OX-2,My033,OX2
Supplier:  ACROBiosystems OX2H5228100UG
Encompass_Preferred
Recombinant Protein;Human CD200 / OX-2 Protein;100UG;Human CD200, His Tag (OX2-H5228) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gly 232 (Accession # AAH22522).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.4 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-101-8369