Learn More
Sino Biological Recombinant Mouse Nogo Receptor/RTN4R Protein (His & hFc Tag)
Supplier:  Sino Biological 50106M03H100
Encompass_Preferred
A DNA sequence encoding the the mature form of mouse RTN4R (NP_075358.1) (Met 1-Ser 447) was fused with the polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1358