Learn More
Hach Company Sension+ 5010T laboratory combination pH electrode for general applications
Catalog No. 501997959 Shop All Hach Company Products
Encompass
Sension+ 5010T is a low maintenance combination pH electrode with a polycarbonate body, non-refillable gel-electrolyte reference and built-in temperature sensor. The 5010T has a fixed 1 meter cable with BNC connector (pH) and banana (temperature) connectors. It is intended for use with Hach Sension+ Laboratory pH meters. The 5010T has a ceramic pin junction and is ideal for pH measurements in general aqueous applications.
Catalog No. 50-199-7959 Supplier Hach Company Supplier No. LZW5010T.97.002
Sension+ 5010T is a low maintenance combination pH electrode with a polycarbonate body, non-refillable gel-electrolyte reference and built-in temperature sensor. The 5010T has a fixed 1 meter cable with BNC connector (pH) and banana (temperature) connectors. It is intended for use with Hach Sension+ Laboratory pH meters. The 5010T has a ceramic pin junction and is ideal for pH measurements in general aqueous applications.