Learn More
Sino Biological Recombinant Mouse CD27/TNFRSF7 Protein (His Tag)
Supplier:  Sino Biological 50110M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Arg 182) of mouse CD27 (NP_001028298.1) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1364