Learn More
Sino Biological Recombinant Mouse IL-1 alpha/IL1A Protein
Supplier:  Sino Biological 50114MNAE100
Encompass_Preferred
A DNA sequence encoding the mature form of mouse IL1A (NP_034684.2) (Ser 115-Ser 270) was expressed, with an initial Met at the C-terminus.
Catalog No. 50-162-1374