Learn More
Sino Biological Recombinant Mouse PGRP-S/PGLYRP1 Protein (His Tag)
Supplier:  Sino Biological 50115M08H100
Encompass_Preferred
A DNA sequence encoding the mouse PGLYRP (O88593) (Met 1-Glu 182) was expressed, with a C-terminal polyhistidine tag
Catalog No. 50-162-1377