Learn More
Sino Biological Recombinant Mouse SIGIRR Protein (His & hFc Tag)
Supplier:  Sino Biological 50122M03H100
Encompass_Preferred
A DNA sequence encoding the mouse SIGIRR (NP_075546.2) (Met 1-His 117) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1389