Learn More
Sino Biological Anti-PD-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50124RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse (rM PDCD1; Catalog#50124-M08H; Met 1-Gln 167; NP_032824.1). PDCD1 specific IgG was purified by mouse specific affinity chromatography.
Catalog No. 50-202-3503