Learn More
Sino Biological Recombinant Mouse AXL Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50126M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Pro 443) of mouse AXL (NP_033491.2) precursor was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1402