Learn More
Hach Company Sension+ pH electrode, for universal use, glass body
Catalog No. 501997961 Shop All Hach Company Products
Encompass
pH liquid electrode (refillable) with 2 ceramic diaphragms and silver ion barrier. Glass body. Range 0-14 pH, -10 to 100 ºC. 1 m cable. For Laboratory Meter.This pH electrode is suitable for aqueous media in general and for samples containing sulphurs, reducing sugars or other substances which react with silver ions.High stability reference electrode due to sleeved cartridge and Ag+ ion barrier design.
Catalog No. 50-199-7961 Supplier Hach Company Supplier No. LZW5014.97.0002
pH liquid electrode (refillable) with 2 ceramic diaphragms and silver ion barrier. Glass body. Range 0-14 pH, -10 to 100 ºC. 1 m cable. For Laboratory Meter.This pH electrode is suitable for aqueous media in general and for samples containing sulphurs, reducing sugars or other substances which react with silver ions.High stability reference electrode due to sleeved cartridge and Ag+ ion barrier design.