Learn More
Sino Biological Recombinant Mouse TPO/Thrombopoietin Protein (His Tag)
Catalog No. 501621446
Encompass_Preferred
A DNA sequence encoding the mouse THPO (NP_033405.1) precursor (Met 1-Thr 356) with a N-terminal polyhistidine tag was expressed.
Catalog No. 50-162-1446 Supplier Sino Biological Supplier No. 50146M08H100
A DNA sequence encoding the mouse THPO (NP_033405.1) precursor (Met 1-Thr 356) with a N-terminal polyhistidine tag was expressed.