Learn More
Sino Biological Recombinant Mouse TROY/TNFRSF19 Protein (His Tag)
Supplier:  Sino Biological 50148M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Leu 170) of mouse TNFRSF19 (NP_001157627.1) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1451