Learn More
Sino Biological Recombinant Mouse TREM-2 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 50149M08H100
Encompass_Preferred
A DNA sequence encoding the mouse TREM2 (NP_112544.1) extracellular domain (Met 1-Pro 168) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-1455