Learn More
Hach Company Sension+ 5014T pH liquid combination electrode, with silver ion barrier
Catalog No. 501997962 Shop All Hach Company Products
Encompass
pH liquid electrode (refillable) with 2 ceramic diaphragms and silver ion barrier. Automatic temperature compensation. Glass body. Range 0-14 pH, -10 to 100 ºC. 1 m cable. For Laboratory Meter.This pH electrode is suitable for aqueous media in general and for samples containing sulphurs, reducing sugars or other substances which react with silver ions.Reference electrode stability due to sleeved cartridge and Ag+ ion barrier design.
Catalog No. 50-199-7962 Supplier Hach Company Supplier No. LZW5014T.97.002
pH liquid electrode (refillable) with 2 ceramic diaphragms and silver ion barrier. Automatic temperature compensation. Glass body. Range 0-14 pH, -10 to 100 ºC. 1 m cable. For Laboratory Meter.This pH electrode is suitable for aqueous media in general and for samples containing sulphurs, reducing sugars or other substances which react with silver ions.Reference electrode stability due to sleeved cartridge and Ag+ ion barrier design.