Learn More
Sino Biological VEGF164 Protein, Mouse, Recombinant, HPLC-verified
Catalog No. 501621473
Encompass_Preferred
A DNA sequence encoding the mouse VEGF164 (isoform VEGF-1) (Q00731-2) (Met 1-Arg 190) was expressed and purified.
Catalog No. 50-162-1473 Supplier Sino Biological Supplier No. 50159MNAB100
Add to Cart
Add to Cart
A DNA sequence encoding the mouse VEGF164 (isoform VEGF-1) (Q00731-2) (Met 1-Arg 190) was expressed and purified.