Learn More
Sino Biological Recombinant Mouse uPAR/PLAUR Protein (His & hFc Tag)
Supplier:  Sino Biological 50160M03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Thr 297) of mouse PLAUR (NP_035243.1) precursor was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1477