Learn More
Sino Biological Recombinant Mouse TLR3 Protein (His Tag)
Supplier:  Sino Biological 50161M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse TLR3 (NP_569054.2) (Met 1-Leu 705) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-1483