Learn More
Sino Biological Recombinant Mouse TWSG1 Protein (His Tag)
Supplier:  Sino Biological 50162M08H100
Encompass_Preferred
A DNA sequence encoding the mouse TWSG1 (Q9EP52) (Met 1-Phe 222) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-1485