Learn More
ACROBiosystems Recombinant Protein;Human Ephrin-A1 / EFNA1 Protein, Fc Tag;HEK293;100UG;EF1,EFNA1,Ephrin-A1,Ephrin A1,LERK-1,LERK1,TNFAIP4,TNFAIP-4,EPLG1,EPLG-1,B61,ECKLG,EFL1
Supplier: ACROBiosystems EF1H5251100UG
Recombinant Protein;Human Ephrin-A1 / EFNA1 Protein, Fc Tag;100UG;Human Ephrin-A1, Fc Tag (EF1-H5251) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Ser 182 (Accession # NP_004419).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46 kDa. As a result of glycosylation, the protein migrates as 50-55 kDa under reducing (R) condition, and 100-110 kDa under non-reducing (NR) condition (SDS-PAGE).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.