Learn More
Sino Biological Recombinant Mouse ACVR2B Protein (His & hFc Tag), HPLC-verified
Supplier:  Sino Biological 50173M03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Thr 134) of mouse ACVR2B (NP_031423.1) precursor was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1503