Learn More
Sino Biological Recombinant Mouse Artemin Protein (hFc Tag)
Supplier:  Sino Biological 50176M01H100
Encompass_Preferred
A DNA sequence encoding the mouse Artn (NP_033841.1) (Ala112-Gly224) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-1513