Learn More
Sino Biological Recombinant Mouse I-309/CCL1 Protein (hFc Tag)
Supplier:  Sino Biological 50178M01H50
Encompass_Preferred
A DNA sequence encoding the secreted form of mouse CCL1 isoform 1 (P10146-1) (Lys 24-Cys 92) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-1519