Learn More
Sino Biological Recombinant Mouse, Rat FGF1 Protein
Supplier:  Sino Biological 50179MNAE10
Encompass_Preferred
A DNA sequence encoding the mature form of mouse / rat FGF1 (NP_034327.1) (Phe16-Asp155) was expressed. Mouse and Rat FGF1 sequences are identical.
Catalog No. 50-162-1521