Learn More
Sino Biological Recombinant Mouse Reg3A Protein (His Tag)
Supplier:  Sino Biological 50185M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Gln175) of mouse REG3A (NP_035389.1) precursor was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-1529