Learn More
Sino Biological Recombinant Mouse VSIG4 Protein (His & hFc Tag)
Supplier:  Sino Biological 50187M03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Pro 187) of mouse VSIG4 (NP_808457.1) precursor was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1535