Learn More
Sino Biological Recombinant Mouse COLEC10 Protein (hFc Tag)
Supplier:  Sino Biological 50189M01H100
Encompass_Preferred
A DNA sequence encoding the mouse COLEC10 (Q8CF98) (Cys119-Lys277) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-1543