Learn More
Sino Biological Anti-CD38 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50191RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD38 (rM CD38; Catalog#50191-M08H; NP_031672.2; Leu 45-Thr 304). CD38 specific IgG was purified by mouse CD38 affinity chromatography.
Catalog No. 50-202-3841