Learn More
Sino Biological Recombinant Mouse VE-Cadherin Protein (His Tag)
Supplier:  Sino Biological 50192M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CDH5 (NP_033998.2) extracellular domain (Met 1-Ala 592) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1552