Learn More
Sino Biological Recombinant Mouse COLEC11 Protein (His Tag)
Supplier:  Sino Biological 50195M08B100
Encompass_Preferred
A DNA sequence encoding the mouse COLEC11 (Q3SXB8-1) (Met1-Leu272) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-1560