Learn More
ACROBiosystems ACROBiosystems Mouse Siglec-15 / CD33L3 Protein, His Tag
Supplier:  ACROBiosystems SG5M52H7100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse Siglec-15, His Tag (SG5-M52H7) is expressed from human 293 cells (HEK293). It contains AA Arg 24 - Thr 262 (Accession # A7E1W8-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.5 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse Siglec-15, His Tag (Cat. No. SG5-M52H7) at 5 μg/mL (100 μL/well) can bind Mouse Human chimeric MAb (5G12, Human IgG1) with a linear range of 0.3-2 ng/mL (QC tested).;Immobilized Mouse Siglec-15, His Tag (Cat. No. SG5-M52H7) at 10 μg/mL (100 μL/well) can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with a linear range of 0.078-1.25 μg/mL (Routinely tested).
Catalog No. 50-195-6897