Learn More
Sino Biological Recombinant Mouse SLAMF7/CRACC/CS1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50201M08H100
Encompass_Preferred
A DNA sequence encoding the mouse SLAMF7 (NP_653122.2) extracellular domain (Met 1-Gly 224) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1564