Learn More
ACROBiosystems Human Siglec-15 / CD33L3 Protein, Mouse IgG2a Fc Tag
Supplier: ACROBiosystems SG5H5254100UG
Recombinant Protein;100UG;Human Siglec-15, Mouse IgG2a Fc Tag (SG5-H5254) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Accession # Q6ZMC9-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 53.4 kDa.