Learn More
Sino Biological Recombinant Mouse IL-18BP Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50206M08H100
Encompass_Preferred
A DNA sequence encoding mouse IL18BP (NP_034661.1) (Met 1-Ala 193) precursor was expressed with the polyhistide tag at the C-terminus.
Catalog No. 50-162-1572