Learn More
New England Biolabs, Inc. Luna SARS-CoV-2 RT-qPCR Multiplex Assay Kit - 96 rxn
Supplier: New England Biolabs, Inc. E3019S
The Luna SARS-CoV-2 RT-qPCR Multiplex Assay Kit is optimized for real-time qualitative detection of SARS-CoV-2 nucleic acid using hydrolysis probes. It features the Luna Probe One-Step RT-qPCR 4X Mix with UDG, an optimized SARS-CoV-2 Primer/Probe mix containing primers and probes specific to two regions of the SARS-CoV-2 virus N-gene, and a positive control template. The probes have been modified to contain different fluorophores (N1, HEX; N2, FAM) to enable simultaneous observation on two different channels of a real-time instrument. To ensure the integrity of the input material and absence of inhibition, an internal control (IC) primer and probe set, designed to amplify the human RNaseP gene, is also provided in the primer mix. The reverse primer of this target has been modified from the CDC design to target an exon/exon boundary to reduce background amplification from possible contaminating genomic DNA. Amplification of the IC is observed in the Cy5 channel.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.