Learn More
Sino Biological Recombinant Mouse CD200R Protein (His & hFc Tag)
Supplier:  Sino Biological 50209M03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Pro 238) of mouse CD200R1 (NP_067300.1) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1579