Learn More
Electron Microscopy Sciences Transfer Pipette; Polypropylene, 50/PK
SDP
Catalog No. 502090959
Encompass_Preferred
It works as a syringe. Inexpensive and Disposable. Good for liquid transfering tasks. Each pipette has calibration markings from 0 to 12ml in 0.2ml increments. Tubing may be attached to the tip to suit your application, and tips may also be cut to facilitate aspiration and delivery of fluids of varying viscosities.
Catalog No. 50-209-0959 Supplier Electron Microscopy Sciences Supplier No. 7098620
It works as a syringe. Inexpensive and Disposable. Good for liquid transfering tasks. Each pipette has calibration markings from 0 to 12ml in 0.2ml increments. Tubing may be attached to the tip to suit your application, and tips may also be cut to facilitate aspiration and delivery of fluids of varying viscosities.