Learn More
New England Biolabs, Inc. NEBNext ARTIC SARS-CoV-2 RT-PCR Module 96 rxn
SDP
Supplier:  New England Biolabs, Inc. E7626L
Encompass
The ARTIC SARS-CoV-2 protocol from the ARTIC Network is a multiplexed amplicon approach covering the whole viral genome. The NEBNext ARTIC SARS-CoV-2 RT-PCR Module is based on this method and incorporates optimized reagents for the cDNA synthesis and targeted cDNA amplification steps. A single RT protocol is used regardless of input amount (10-10,000 viral genome copies), and no normalization step is required ahead of targeted amplification. The multiplex primers are balanced to provide uniform amplicon coverage across the viral genome.
Catalog No. 50-210-8559