Learn More
Sino Biological Recombinant Mouse IL-36RN/IL-1F5 Protein
Supplier:  Sino Biological 50213MNAE50
Encompass_Preferred
A DNA sequence encoding the mouse IL1F5 (NP_062324.2) (Val 3-Asp 156) was expressed and purified..
Catalog No. 50-162-1584