Learn More
Sino Biological Recombinant Mouse PILR-beta Protein (hFc Tag)
Supplier:  Sino Biological 50219M02H100
Encompass_Preferred
A DNA sequence encoding the mouse PILRB1 (Q2YFS2) extracellular domain (Met 1-Gly 193) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1593