Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Labs 3.5 - 5 kD Biotech CE Dialysis Kits
SDP

Catalog No. 08801249 Shop All Spectrum Medical Industries Products
Change view
Click to view available options
Diameter (Metric):
6.4 mm
10 mm
15 mm
20 mm
Length (Metric):
0.32 mL/cm vol./length
0.79 mL/cm vol./length
1.8 mL/cm vol./length
3.1 mL/cm vol./length
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric)
08-801-249 10 mm 0.79 mL/cm vol./length
08-700-120 20 mm 3.1 mL/cm vol./length
08-700-118 6.4 mm 0.32 mL/cm vol./length
08-700-119 15 mm 1.8 mL/cm vol./length
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 08-801-249 Supplier Spectrum™ Supplier No. 131198T
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • 1m Biotech CE Tubing in 0.05% sodium azide
  • Two universal tubing closures
  • Five opening picks

With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane

Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC

  • Hydrophilic, symmetric porositiy
  • For small tubing demand applications or evaluating a specific dialysis membrane
  • Includes 1m tubing with pair of tubing closures and 5 opening picks

Specifications

Product Type Dialysis Trial Kit
MWCO 3.5 to 5 kDa
Material Cellulose Ester
Format Tubing
Diameter (Metric) 10 mm
Diameter (English) 0.39 in.
Length (Metric) 0.79 mL/cm vol./length
Kit Contents 1m roll Biotech grade CE dialysis membrane tubing, 2 universal closures, 5 opening picks
Quantity Each
For Use With (Application) Ideal for small, critical dialysis applications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.