Learn More
Sino Biological Recombinant Mouse S100A15 Protein (His & MBP Tag)
Supplier:  Sino Biological 50226M10E50
Encompass_Preferred
A DNA sequence encoding the mouse S100A7A (Q6S5I3) (Met 1-Tyr 108) was fused with the N-terminal polyhistidine-tagged MBP tag at the N-terminus.
Catalog No. 50-162-1600