Learn More
Sino Biological Recombinant Mouse DNAM-1/CD226 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50232M08H100
Encompass_Preferred
A DNA sequence encoding the mouse DNAM1 (NP_848802.2) extracellular domain (Met 1-Pro 254) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1611