Learn More
Sino Biological Recombinant Mouse PEDF/Serpin F1 Protein (His Tag)
Supplier:  Sino Biological 50235M08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Thr 417) of mouse SerpinF1 (NP_035470.3) precursor was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-1618