Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Labs Spectra/Por™ 1 6-8 kD MWCO Standard RC Dry Dialysis Kits
SDP

Catalog No. 08700148 Shop All Spectrum Medical Industries Products
Change view
Click to view available options
Diameter (Metric):
6.4 mm
14.6 mm
20.4 mm
25.5 mm
32 mm
64 mm
76 mm
Length (Metric):
0.32 mL/cm vol./length
1.7 mL/cm
32 mL/cm vol./length
46 mL/cm vol./length
5 m
5.1 mL/cm vol./length
Quantity:
1 m Tubing/Kit
5 m Tubing/Kit
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric) Quantity
08-700-148 76 mm 46 mL/cm vol./length 5 m Tubing/Kit
08-700-146 64 mm 32 mL/cm vol./length 5 m Tubing/Kit
08-700-143 25.5 mm 5.1 mL/cm vol./length 5 m Tubing/Kit
08-700-140 14.6 mm 1.7 mL/cm 5 m Tubing/Kit
08-700-138 6.4 mm 0.32 mL/cm vol./length 5 m Tubing/Kit
08-801-241 20.4 mm 5 m Each
08-801-242 32 mm 5 m Each
08-700-147 64 mm 32 mL/cm vol./length 1 m Tubing/Kit
08-700-144 25.5 mm 5.1 mL/cm vol./length 1 m Tubing/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 08-700-148 Supplier Spectrum™ Supplier No. 132675T
Add to Cart
Add to Cart

Includes

  • 1m or 5m of Spectra/Por 1 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers

With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated

  • For basic dialysis applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water)

Specifications

MWCO 6 to 8 kDa
Diameter (Metric) 76 mm
Length (Metric) 46 mL/cm vol./length
Quantity 5 m Tubing/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.