Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, 1/32 in by 3/32 in, 50 ft
Supplier:  Accuflow Systems ACF00001
Encompass_Preferred
Crystal-clear, phthalate-free laboratory tubing designed for instrumentation connections, venting, drain lines, and vacuum applications. The tubing resists flex-fatigue and abrasion, features a glassy-smooth inner bore to minimize buildup, and is marked at 1-foot intervals for easy measurement. Suitable for autoclave and ethylene oxide sterilization, supplied in 50 ft lengths with 1/32 in I.D. and 3/32 in O.D.
  • Crystal-clear, phthalate-free construction.
  • Resists flex-fatigue and abrasion.
  • Marked at 1-foot intervals for measurement.
  • Glassy-smooth inner bore reduces buildup.
  • Sterilizable by autoclave and ethylene oxide.
  • Dimensions: 1/32 in I.D., 3/32 in O.D., 50 ft length.
Catalog No. 50-236-5907