Learn More
Teknova 20X TBS with 1% Tween-20, pH 7.5. (2M Tris, 3M NaCl) 1000mL, Sterile. 2 pack.
SDP
Supplier:  Teknova T16882PK
Encompass
2M Tris,3M Sodium Chloride,1.0% Tween-20
Catalog No. 50-238-7352