Learn More
Teknova Standard Methods Agar (Plate Count Agar). 100mm, 20 Plates per Sleeve, Sterile.
SDP
Supplier:  Teknova S0144
Encompass
0.5% Tryptone, 0.25% Yeast Extract,0.1% Cerelose Anhydrous Dextrose (Glucose), 1.5% Agar
Catalog No. 50-238-8059