Learn More
Teknova 10mM Tris-HCl, pH 8.0. 2mL Tubes, 100-Pack, Sterile. DNase/RNase Tested.
SDP
Supplier:  Teknova T1180
Encompass
Tris (tris(hydroxymethyl)aminomethane) is used as a component ofbuffer solutions, especially for solutions ofnucleic acids. The useful buffer range for tris (pH 7–9) is similar to the pH of most living organisms. This, and its low cost, make tris one of the most common buffers in the biology/biochemistry laboratory.
Catalog No. 50-238-8073